Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046542.2 | NP_001040007.1 | 10 - 11 | 1120 | 93 | |
GenBank mRNA | BC112463.1 | - | 10 - 11 | 1121 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |