Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC22C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046546.2 | NP_001040011.1 | 2 - 3 | 376 | 55 | |
GenBank mRNA | BC110195.1 | - | 2 - 3 | 377 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |