Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046566.2 | NP_001040031.1 | 2 - 3 | 166 | 108 | |
GenBank mRNA | BC105369.1 | - | 2 - 3 | 167 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |