Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046612.1 | NP_001040077.1 | 5 - 6 | 645 | 108 | |
GenBank mRNA | BC112763.1 | - | 5 - 6 | 645 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |