Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075604.2 | NP_001069072.2 | 6 - 7 | 472 | 142 | |
NR_045536.1 | 6 - 7 | 613 | 142 | |||
GenBank mRNA | BC110259.1 | - | 6 - 7 | 614 | 142 | |
DT858178.1 | - | 6 - 7 | 473 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |