Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075833.2 | NP_001069301.1 | 5 - 6 | 551 | 89 | |
GenBank mRNA | BC110215.1 | - | 5 - 6 | 552 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |