Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075894.1 | NP_001069362.1 | 1 - 2 | 497 | 112 | |
GenBank mRNA | BC105392.1 | - | 1 - 2 | 497 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |