Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRGC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076146.2 | NP_001069614.1 | 1 - 2 | 33 | 109 | |
GenBank mRNA | BC109879.1 | - | 1 - 2 | 34 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |