Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076438.2 | NP_001069906.1 | 1 - 2 | 278 | 99 | |
GenBank mRNA | BC109690.1 | - | 1 - 2 | 280 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |