Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYRM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076505.2 | NP_001069973.1 | 1 - 2 | 43 | 88 | |
GenBank mRNA | BC111663.1 | - | 1 - 2 | 44 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |