Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015561.2 | NP_001015561.1 | 2 - 3 | 315 | 71 | |
GenBank mRNA | AY911381.1 | - | 3 - 4 | 335 | 71 | |
BC113281.1 | - | 2 - 3 | 315 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |