Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034422.2 | NP_001029594.1 | 5 - 6 | 648 | 138 | |
GenBank mRNA | BC118201.1 | - | 5 - 6 | 648 | 138 | |
BT021696.1 | - | 5 - 6 | 629 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |