Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045873.1 | NP_001039338.1 | 3 - 4 | 411 | 115 | |
GenBank mRNA | BC114065.1 | - | 3 - 4 | 411 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |