Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045905.2 | NP_001039370.1 | 6 - 7 | 685 | 71 | |
GenBank mRNA | BC114867.1 | - | 6 - 7 | 686 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |