Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDNEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046026.1 | NP_001039491.1 | 3 - 4 | 631 | 67 | |
GenBank mRNA | BC114677.1 | - | 3 - 4 | 631 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |