Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046147.1 | NP_001039612.1 | 6 - 7 | 545 | 78 | |
GenBank mRNA | BC113324.1 | - | 6 - 7 | 545 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |