Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075149.2 | NP_001068617.1 | 1 - 2 | 201 | 69 | |
GenBank mRNA | BC114816.1 | - | 1 - 2 | 202 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |