Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075446.2 | NP_001068914.1 | 4 - 5 | 576 | 85 | |
GenBank mRNA | AB238940.1 | - | 2 - 3 | 361 | 85 | |
BC118359.1 | - | 4 - 5 | 578 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |