Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5C3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075515.2 | NP_001068983.2 | 5 - 6 | 374 | 98 | |
GenBank mRNA | BC118394.1 | - | 5 - 6 | 350 | 98 | |
EE241538.1 | - | 5 - 6 | 375 | 98 | ||
EH136301.1 | - | 5 - 6 | 370 | 98 | ||
EH208684.1 | - | 5 - 6 | 339 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |