Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A6OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075567.1 | NP_001069035.1 | 1 - 2 | 199 | 131 | |
GenBank mRNA | BC116007.1 | - | 1 - 2 | 199 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |