Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBOX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075669.1 | NP_001069137.1 | 6 - 7 | 1640 | 100 | |
GenBank mRNA | BC114651.1 | - | 6 - 7 | 1640 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |