Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C25H7orf26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075754.2 | NP_001069222.1 | 3 - 4 | 592 | 69 | |
GenBank mRNA | BC118367.1 | - | 3 - 4 | 594 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |