Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046506.1 | NP_001039971.1 | 2 - 3 | 231 | 101 | |
GenBank mRNA | BC122611.1 | - | 2 - 3 | 231 | 101 | |
DQ335873.1 | - | 2 - 3 | 262 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |