Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075645.1 | NP_001069113.1 | 3 - 4 | 288 | 68 | |
GenBank mRNA | BC119960.1 | - | 3 - 4 | 288 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |