Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075663.1 | NP_001069131.1 | 8 - 9 | 1325 | 92 | |
GenBank mRNA | BT026277.1 | - | 8 - 9 | 1325 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |