Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IP6K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076010.1 | NP_001069478.1 | 2 - 3 | 570 | 93 | |
GenBank mRNA | BC122746.1 | - | 2 - 3 | 570 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |