Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAFA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076435.1 | NP_001069903.1 | 4 - 5 | 518 | 85 | |
GenBank mRNA | BC120098.1 | - | 4 - 5 | 518 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |