Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076805.1 | NP_001070273.1 | 7 - 8 | 1389 | 92 | |
GenBank mRNA | BC123887.1 | - | 7 - 8 | 1389 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |