Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037141.3 | NP_001032218.2 | 3 - 4 | 313 | 59 | |
NM_001110194.1 | NP_001103664.1 | 6 - 7 | 714 | 59 | ||
GenBank mRNA | DQ219802.1 | - | 3 - 4 | 315 | 59 | |
DT721910.1 | - | 2 - 3 | 173 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |