Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077020.1 | NP_001070488.1 | 3 - 4 | 275 | 142 | |
GenBank mRNA | BC105572.1 | - | 3 - 4 | 291 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |