Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCGF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077980.1 | NP_001071448.1 | 2 - 3 | 353 | 70 | |
GenBank mRNA | BC126565.1 | - | 2 - 3 | 353 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |