Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCGF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077980.1 | NP_001071448.1 | 5 - 6 | 556 | 144 | |
GenBank mRNA | BC126565.1 | - | 5 - 6 | 556 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |