Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078025.1 | NP_001071493.1 | 2 - 3 | 1064 | 59 | |
GenBank mRNA | BC126844.1 | - | 2 - 3 | 1064 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |