Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079775.1 | NP_001073243.1 | 1 - 2 | 488 | 148 | |
GenBank mRNA | BC126761.1 | - | 1 - 2 | 488 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |