Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080332.2 | NP_001073801.1 | 2 - 3 | 181 | 95 | |
GenBank mRNA | BC126836.1 | - | 2 - 3 | 182 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |