Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081546.1 | NP_001075015.1 | 1 - 2 | 72 | 92 | |
GenBank mRNA | BC122626.1 | - | 1 - 2 | 72 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |