Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083459.2 | NP_001076928.1 | 1 | 127 | 68 | |
GenBank mRNA | BC114915.1 | - | 1 | 129 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Both primers and probe map within a single exon |