Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101045.1 | NP_001094515.1 | 3 - 4 | 310 | 149 | |
GenBank mRNA | BC151457.1 | - | 3 - 4 | 310 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |