Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173901.3 | NP_776326.1 | 2 - 3 | 204 | 99 | |
GenBank mRNA | X00050.1 | - | 176 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |