Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024473.2 | NP_001019644.2 | 5 - 6 | 804 | 121 | |
GenBank mRNA | BC114679.1 | - | 5 - 6 | 804 | 121 | |
BT021780.1 | - | 5 - 6 | 868 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |