Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCT6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034542.2 | NP_001029714.1 | 7 - 8 | 955 | 94 | |
GenBank mRNA | BC105191.1 | - | 7 - 8 | 953 | 94 | |
BC142274.1 | - | 7 - 8 | 937 | 94 | ||
BT030538.1 | - | 7 - 8 | 955 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |