Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TULP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098873.2 | NP_001092343.1 | 4 - 5 | 434 | 63 | |
GenBank mRNA | BC146115.1 | - | 4 - 5 | 435 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |