Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098965.1 | NP_001092435.1 | 1 - 2 | 63 | 73 | |
GenBank mRNA | BC134798.1 | - | 1 - 2 | 64 | 73 | |
BC142035.1 | - | 1 - 2 | 63 | 73 | ||
BC151506.1 | - | 1 - 2 | 64 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |