Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098965.1 | NP_001092435.1 | 3 - 4 | 255 | 60 | |
GenBank mRNA | BC134798.1 | - | 3 - 4 | 256 | 60 | |
BC142035.1 | - | 3 - 4 | 255 | 60 | ||
BC151506.1 | - | 3 - 4 | 256 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |