Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAFG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098980.1 | NP_001092450.1 | 1 - 2 | 219 | 142 | |
GenBank mRNA | BC142248.1 | - | 1 - 2 | 219 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |