Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAPLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098981.2 | NP_001092451.1 | 3 - 4 | 586 | 92 | |
GenBank mRNA | BC142409.1 | - | 3 - 4 | 587 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |