Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098986.1 | NP_001092456.1 | 3 - 4 | 288 | 78 | |
GenBank mRNA | BC142382.1 | - | 3 - 4 | 288 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |