Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100320.1 | NP_001093790.1 | 7 - 8 | 745 | 136 | |
GenBank mRNA | BT030696.1 | - | 7 - 8 | 745 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |