Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101053.2 | NP_001094523.1 | 1 - 2 | 226 | 71 | |
GenBank mRNA | BC151253.1 | - | 1 - 2 | 232 | 71 | |
BC151550.1 | - | 1 - 2 | 227 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |