Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101879.2 | NP_001095349.1 | 3 - 4 | 562 | 84 | |
GenBank mRNA | BC149341.1 | - | 3 - 4 | 563 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |